4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid

OR3786-1G
APOSOR3786-1GEA 30.6 EUR
APOSOR3786-1G
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure 98%
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₃BClNO₃
Molecular Weight: 289,53 g/mol
Schmelzpunkt: 217…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027211
CAS-Nummer: 913835-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR