4-(4-Bromphenylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-((4-Bromophenyl)carbamoyl)phenyl)boronic acid, 4-(4-Bromophenylcarbamoyl)benzeneboronic acid
OR3787-1G
APOSOR3787-1GEA
21.9
EUR
APOSOR3787-1G
4-(4-Bromphenylcarbamoyl)phenylboronsäure 98%
4-(4-Bromphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BBrNO₃ Molecular Weight: 319,95 g/mol Schmelzpunkt: 268…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027201 CAS-Nummer: 874287-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...