4-Brom-2,5-xylidin 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2,5-dimethylaniline

OR3788-1G
APOSOR3788-1GEA 65.4 EUR
APOSOR3788-1G
4-Brom-2,5-xylidin 96%
4-Brom-2,5-xylidin
Formel: C₈H₁₀BrN
Molecular Weight: 200.0757 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07780642
CAS-Nummer: 30273-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR