4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

OR3793-1G
APOSOR3793-1GEA 21.9 EUR
APOSOR3793-1G
4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure 98%
4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD09027212
CAS-Nummer: 913835-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR