4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((2,5-Dimethylphenyl)carbamoyl)phenyl)boronic acid

OR3794-1G
APOSOR3794-1GEA 30.6 EUR
APOSOR3794-1G
4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure 96%
4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
Molecular Weight: 269,1 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09027214
CAS-Nummer: 913835-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR