3-(Piperdin-1-ylmethyl)benzoic acid hydrochloride hemihydrate 97%

Lieferant: Apollo Scientific
Achtung

OR3799-250MG
APOSOR3799-250MGEA 186 EUR
APOSOR3799-250MG
3-(Piperdin-1-ylmethyl)benzoic acid hydrochloride hemihydrate 97%
3-(Piperdin-1-ylmethyl)benzoic acid hydrochloride hemihydrate
Schmelzpunkt: 234…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772860
CAS-Nummer: 863991-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR