1-Methyl-1H-indazole-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Formyl-1-methyl-1H-indazole, 1-Methyl-1H-indazole-3-carbaldehyde

OR3801-250MG
APOSOR3801-250MGEA 49.3 EUR
APOSOR3801-250MG
1-Methyl-1H-indazole-3-carboxaldehyde 97%
1-Methyl-1H-indazole-3-carboxaldehyde
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060478
CAS-Nummer: 4002-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR