3-Morpholin-4-ylmethyl-benzaldehyde 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Morpholinomethyl)benzaldehyde

OR3804-250MG
APOSOR3804-250MGEA 0 EUR
APOSOR3804-250MG
3-Morpholin-4-ylmethyl-benzaldehyde 95+%
3-Morpholin-4-ylmethylbenzaldehyde
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205.26 g/mol
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07782413
CAS-Nummer: 446866-83-7
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR