5-Chlor-2-(methylsulfanyl)-4-pyrimidincarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

OR3810-1G
APOSOR3810-1GEA 21.9 EUR
APOSOR3810-1G
5-Chlor-2-(methylsulfanyl)-4-pyrimidincarbonsäure 98%
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid
Formel: C₆H₅ClN₂O₂S
Molecular Weight: 204,6300 g/mol
Schmelzpunkt: 204…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173907
CAS-Nummer: 61727-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR