5-Acetyl-2-amino-4-methylpyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Amino-4-methylpyrimidin-5-yl)ethanone

OR3847-1G
APOSOR3847-1GEA 23.2 EUR
APOSOR3847-1G
5-Acetyl-2-amino-4-methylpyrimidine
5-Acetyl-2-amino-4-methylpyrimidine
Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
Schmelzpunkt: 226…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052621
CAS-Nummer: 66373-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR