4-(3-Iodobenzyl)morpholine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Iodo-3-[(morpholin-4-yl)methyl]benzene
OR3859-1G
APOSOR3859-1GEA
528
EUR
APOSOR3859-1G
4-(3-Iodobenzyl)morpholine
4-(3-Iodobenzyl)morpholine
Formel:
C₁₁H₁₄INO Molecular Weight: 303,139 g/mol Schmelzpunkt: 32…34 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06797855 CAS-Nummer: 731812-03-6 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...