2,5-Di-tert-butylhydrochinon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
BHQ
OR3870-250G
APOSOR3870-250GEA
29
EUR
APOSOR3870-250G
2,5-Di-tert-butylhydrochinon
2,5-Di-tert-butylhydrochinon
Formel:
C₁₄H₂₂O₂ Schmelzpunkt: 217…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008825 CAS-Nummer: 88-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...