3-[1,2,4]Triazol-1-ylmethylphenylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[(1H-1,2,4-Triazol-1-yl)methyl]aniline

OR3877-1G
APOSOR3877-1GEA 109 EUR
APOSOR3877-1G
3-[1,2,4]Triazol-1-ylmethylphenylamine 97%
3-[1,2,4]Triazol-1-ylmethylphenylamine
Formel: C₉H₁₀N₄
Molecular Weight: 174,21 g/mol
Schmelzpunkt: 85,5…86.5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08060528
CAS-Nummer: 127988-22-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR