3-Cyclopropyl-1H-pyrazol-5-amine 96%

Lieferant: Apollo Scientific
Achtung

OR3887-250MG
APOSOR3887-250MGEA 0 EUR
APOSOR3887-250MG
3-Cyclopropyl-1H-pyrazol-5-amine 96%
3-Cyclopropyl-1H-pyrazol-5-amine
Formel: C₆H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00067985
CAS-Nummer: 175137-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR