2-[(3-Nitro-2-pyridyl)thio]acetic acid

Lieferant: Apollo Scientific

OR3908-1G
APOSOR3908-1GEA 510 EUR
APOSOR3908-1G
2-[(3-Nitro-2-pyridyl)thio]acetic acid
2-[(3-Nitro-2-pyridyl)thio]acetic acid
Formel: C₇H₆N₂O₄S
Molecular Weight: 214,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00174245
CAS-Nummer: 18504-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR