5-(Phenylethynyl)thiophene-2-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Phenylethynyl)thiophene-2-carbaldehyde, 2-Formyl-5-(phenylethynyl)thiophene, 5-(Phenylethynyl)-2-thenaldehyde

OR3911-5G
APOSOR3911-5GEA 0 EUR
APOSOR3911-5G
5-(Phenylethynyl)thiophene-2-carboxaldehyde 97%
5-(Phenylethynyl)thiophene-2-carboxaldehyde
Formel: C₁₃H₈OS
Molecular Weight: 212,267 g/mol
Schmelzpunkt: 91 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00126374
CAS-Nummer: 17257-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR