1-(2-Chlorphenyl)biguanid Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
OR3912-1G
OR3912-25G
OR3912-5G
APOSOR3912-1GEA
27.7
EUR
APOSOR3912-1G
APOSOR3912-25G
APOSOR3912-5G
1-(2-Chlorphenyl)biguanid Hydrochlorid
1-(2-Chlorphenyl)biguanid Hydrochlorid
Formel:
C₈H₁₁Cl₂N₅ Schmelzpunkt: 129…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053016 CAS-Nummer: 19579-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...