1-(2-Chlorphenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR3912-1G OR3912-25G OR3912-5G
APOSOR3912-1GEA 27.7 EUR
APOSOR3912-1G APOSOR3912-25G APOSOR3912-5G
1-(2-Chlorphenyl)biguanid Hydrochlorid
1-(2-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Schmelzpunkt: 129…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053016
CAS-Nummer: 19579-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR