2-Brom-5-chlor-3-methylbenzo[b]thiophen 97%

Lieferant: Apollo Scientific
Achtung

OR3913-1G
APOSOR3913-1GEA 27.7 EUR
APOSOR3913-1G
2-Brom-5-chlor-3-methylbenzo[b]thiophen 97%
2-Brom-5-chlor-3-methylbenzo[b]thiophen
Formel: C₉H₆BrClS
Molecular Weight: 261.57 g/mol
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126363
CAS-Nummer: 175203-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR