5-Chlorobenzofurazan-4-sulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride

OR3923-1G OR3923-250MG
APOSOR3923-1GEA 87 EUR
APOSOR3923-1G APOSOR3923-250MG
5-Chlorobenzofurazan-4-sulphonyl chloride 97%
5-Chlorobenzofurazan-4-sulphonyl chloride
Formel: C₆H₂Cl₂N₂O₃S
Molecular Weight: 253.06 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203878
CAS-Nummer: 175203-78-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR