Ethyl-3-[(2-furylmethyl)amino]propanoate 97%
Lieferant: Apollo Scientific
OR3932-1G
APOSOR3932-1GEA
0
EUR
APOSOR3932-1G
Ethyl-3-[(2-furylmethyl)amino]propanoate 97%
Ethyl-3-[(2-furylmethyl)amino]propanoate
Formel:
C₁₀H₁₅NO₃ Molecular Weight: 197,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00219838 CAS-Nummer: 175203-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...