2,4-Dinitrobenzaldoxime

Lieferant: Apollo Scientific
Achtung

OR3936-1G OR3936-5G
APOSOR3936-1GEA 26.1 EUR
APOSOR3936-1G APOSOR3936-5G
2,4-Dinitrobenzaldoxime
2,4-Dinitrobenzaldoxime
Formel: C₇H₅N₃O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD01070627
CAS-Nummer: 3236-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR