4-(3-Hydroxypropyl)benzeneboronic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: [4-(3-Hydroxypropyl)phenyl]boronic acid

OR3943-1G
APOSOR3943-1GEA 235 EUR
APOSOR3943-1G
4-(3-Hydroxypropyl)benzeneboronic acid 96%
4-(3-Hydroxypropyl)benzeneboronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
Schmelzpunkt: 70…74 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04973734
CAS-Nummer: 850568-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR