2-Methoxybenzeneboronic acid, propanediol cyclic ester 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Methoxyphenyl)-1,3,2-dioxaborinane

OR3951-5G
APOSOR3951-5GEA 84.1 EUR
APOSOR3951-5G
2-Methoxybenzeneboronic acid, propanediol cyclic ester 97%
2-Methoxybenzeneboronic acid, propanediol cyclic ester
Formel: C₁₀H₁₃BO₃
Molecular Weight: 192.019 g/mol
Schmelzpunkt: 56…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659934
CAS-Nummer: 141522-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR