4-(Isopropylsulfonyl)phenylboronsäure 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Isopropylsulphonyl)benzeneboronic acid

OR3968-1G
APOSOR3968-1GEA 21.9 EUR
APOSOR3968-1G
4-(Isopropylsulfonyl)phenylboronsäure 97+%
4-(Isopropylsulfonyl)phenylboronsäure
Formel: C₉H₁₃BO₄S
Molecular Weight: 228,0700 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659866
CAS-Nummer: 850567-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR