4-Phenoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

OR3970-1G
APOSOR3970-1GEA 21.9 EUR
APOSOR3970-1G
4-Phenoxyphenylboronsäure 98%
4-Phenoxyphenylboronsäure
Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR