3-(Butylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Butylaminocarbonyl)phenylboronic acid, 3-(n-Butylcarbamoyl)benzeneboronic acid, (3-(Butylcarbamoyl)phenyl)boronic acid

OR3980-5G
APOSOR3980-5GEA 156 EUR
APOSOR3980-5G
3-(Butylcarbamoyl)phenylboronsäure 97%
3-(Butylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221.06 g/mol
Schmelzpunkt: 216…224 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115691
CAS-Nummer: 397843-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR