(4-Nitrophenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitrophenylacetic acid
OR3989-100G
OR3989-25G
OR3989-1G
APOSOR3989-100GEA
0
EUR
APOSOR3989-100G
APOSOR3989-25G
APOSOR3989-1G
(4-Nitrophenyl)essigsäure 97%
(4-Nitrophenyl)essigsäure
Formel:
(C₆H₄(NO₂))CH₂COOH Schmelzpunkt: 150…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007383 CAS-Nummer: 104-03-0 EINECS: 203-168-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...