(4-Nitrophenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitrophenylacetic acid

OR3989-100G OR3989-25G OR3989-1G
APOSOR3989-100GEA 0 EUR
APOSOR3989-100G APOSOR3989-25G APOSOR3989-1G
(4-Nitrophenyl)essigsäure 97%
(4-Nitrophenyl)essigsäure
Formel: (C₆H₄(NO₂))CH₂COOH
Schmelzpunkt: 150…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007383
CAS-Nummer: 104-03-0
EINECS: 203-168-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR