2-Bromo-3-chlorophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR399001-5G
APOSOR399001-5GEA
137
EUR
APOSOR399001-5G
2-Bromo-3-chlorophenol
2-Bromo-3-chlorophenol
Formel:
C₆H₄BrClO Molecular Weight: 207.452 g/mol Siedepunkt: 225 °C Schmelzpunkt: 55.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037216 CAS-Nummer: 855836-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...