2-Brom-5-methoxyphenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-2-hydroxy-4-methoxybenzol, 4-Brom-3-hydroxyanisol

OR399007-5G
APOSOR399007-5GEA 75.4 EUR
APOSOR399007-5G
2-Brom-5-methoxyphenol 98%
2-Brom-5-methoxyphenol
Formel: C₇H₇BrO₂
Lagertemperatur: Kühlschrank
MDL: MFCD03428523
CAS-Nummer: 63604-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR