3-Bromo-4-methoxyphenol 98%

Lieferant: Apollo Scientific
Achtung

OR399010-25G
APOSOR399010-25GEA 185 EUR
APOSOR399010-25G
3-Bromo-4-methoxyphenol 98%
3-Bromo-4-methoxyphenol
Formel: C7H7BrO2
Molecular Weight: 203.0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054143
CAS-Nummer: 17332-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR