3-Chloro-5-methoxyphenol

Lieferant: Apollo Scientific

OR399015-5G
APOSOR399015-5GEA 165 EUR
APOSOR399015-5G
3-Chloro-5-methoxyphenol
3-Chloro-5-methoxyphenol
Formel: C₇H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002260
CAS-Nummer: 65262-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR