3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid

Lieferant: Apollo Scientific
Achtung

OR3995-250MG
APOSOR3995-250MGEA 50.9 EUR
APOSOR3995-250MG
3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid
3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid
Formel: C₁₃H₁₂N₂O₅
Schmelzpunkt: 206…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203067
CAS-Nummer: 387350-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR