3-(Piperidin-1-carbonyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR3998-1G
APOSOR3998-1GEA 33.5 EUR
APOSOR3998-1G
3-(Piperidin-1-carbonyl)phenylboronsäure 97%
3-(Piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₆BNO₃
Molecular Weight: 233,0713 g/mol
Schmelzpunkt: 150…154 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115686
CAS-Nummer: 850568-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR