3-(4-Ethoxybenzoyl)acrylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR4000-1G
APOSOR4000-1GEA 139 EUR
APOSOR4000-1G
3-(4-Ethoxybenzoyl)acrylic acid 98%
3-(4-Ethoxybenzoyl)acrylic acid
Formel: C₁₂H₁₂O₄
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075337
CAS-Nummer: 29582-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR