4-Chlor-3-pyridinboronsäure 95%

Lieferant: Apollo Scientific
Achtung

OR40000-250MG
APOSOR40000-250MGEA 509 EUR
APOSOR40000-250MG
4-Chlor-3-pyridinboronsäure 95%
4-Chloro-3-pyridineboronic acid
Formel: C₅H₅BClNO₂
Molecular Weight: 157,36 g/mol
Schmelzpunkt: 69,3…69.7 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06801679
CAS-Nummer: 452972-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR