6-Chloroisatoic anhydride

Lieferant: Apollo Scientific
Achtung

OR400007-1G OR400007-5G OR400007-25G
APOSOR400007-1GEA 0 EUR
APOSOR400007-1G APOSOR400007-5G APOSOR400007-25G
6-Chloroisatoic anhydride
6-Chloroisatoic anhydride
Formel: C₈H₄ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09032504
CAS-Nummer: 20829-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR