5-Methoxyisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
OR400011-250MG
APOSOR400011-250MGEA
0
EUR
APOSOR400011-250MG
5-Methoxyisatoic anhydride
5-Methoxyisatoic anhydride
Formel:
C₉H₇NO₄ Schmelzpunkt: 244…246 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179411 CAS-Nummer: 37795-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...