5-Methylisatosäureanhydrid

Lieferant: Apollo Scientific

Synonyme: 6-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

OR400012-5G
APOSOR400012-5GEA 98.6 EUR
APOSOR400012-5G
5-Methylisatosäureanhydrid
5-Methylisatosäureanhydrid
Formel: C₉H₇NO₃
Molecular Weight: 177,1568 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809783
CAS-Nummer: 4692-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR