4-Chlorisatosäureanhydrid

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

OR400016-25G
APOSOR400016-25GEA 120 EUR
APOSOR400016-25G
4-Chlorisatosäureanhydrid
4-Chlorisatosäureanhydrid
Formel: C₈H₄ClNO₃
Molecular Weight: 197.58 g/mol
Schmelzpunkt: 255…259 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00090431
CAS-Nummer: 40928-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR