3-(3,4,5-Trimethoxyphenyl)-1-propene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Allyl-1,2,3-trimethoxybenzene
OR40002-1G
APOSOR40002-1GEA
105
EUR
APOSOR40002-1G
3-(3,4,5-Trimethoxyphenyl)-1-propene
3-(3,4,5-Trimethoxyphenyl)-1-propene
Formel:
C₁₂H₁₆O₃ Molecular Weight: 208,26 g/mol Siedepunkt: 106…108 °C/1mmHg Dichte: 1,07 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01656688 CAS-Nummer: 487-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...