4,5-Dimethoxyisatosäureanhydrid

Lieferant: Apollo Scientific

Synonyme: 6,7-Dimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione

OR400025-5G
APOSOR400025-5GEA 84.1 EUR
APOSOR400025-5G
4,5-Dimethoxyisatosäureanhydrid
4,5-Dimethoxyisatosäureanhydrid
Formel: C₁₀H₉NO₅
Molecular Weight: 223,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426390
CAS-Nummer: 20197-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR