3-Acetylbenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone
OR40003-1G
APOSOR40003-1GEA
667
EUR
APOSOR40003-1G
3-Acetylbenzophenon
3-Acetylbenzophenon
Formel:
C₁₅H₁₂O₂ Molecular Weight: 224,2546 g/mol Siedepunkt: 80…85 °C/0,1mmHg Dichte: 1,22 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01662280 CAS-Nummer: 66067-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...