2-Bromo-4-chlorobenzyl bromide 95%

Lieferant: Apollo Scientific

OR400062-25G OR400062-5G
APOSOR400062-25GEA 0 EUR
APOSOR400062-25G APOSOR400062-5G
2-Bromo-4-chlorobenzyl bromide 95%
2-Bromo-4-chlorobenzyl bromide
Formel: C₇H₅Br₂Cl
Lagertemperatur: Raumtemperatur
MDL: MFCD12024978
CAS-Nummer: 33924-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR