2-Bromo-4-chlorophenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-4-chlorophenyl)acetonitrile

OR400067-250MG
APOSOR400067-250MGEA 92.8 EUR
APOSOR400067-250MG
2-Bromo-4-chlorophenylacetonitrile 97%
2-Bromo-4-chlorophenylacetonitrile
Formel: C₈H₅BrClN
Molecular Weight: 230,489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044834
CAS-Nummer: 52864-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR