6-Hydroxyquinazolin-4(1H)-one

Lieferant: Apollo Scientific

Synonyme: 6-Hydroxyquinazolin-4-ol, 6-Hydroxy-4-quinazolone

OR400075-1G OR400075-25G OR400075-5G
APOSOR400075-1GEA 78.3 EUR
APOSOR400075-1G APOSOR400075-25G APOSOR400075-5G
6-Hydroxyquinazolin-4(1H)-one
6-Hydroxyquinazolin-4(1H)-one
Formel: C₈H₆N₂O₂
Molecular Weight: 162.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662723
CAS-Nummer: 16064-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR