4-Bromo-3-iodobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR400078-250MG OR400078-1G
APOSOR400078-250MGEA 87 EUR
APOSOR400078-250MG APOSOR400078-1G
4-Bromo-3-iodobenzyl bromide
4-Bromo-3-iodobenzyl bromide
Formel: C₇H₅Br₂I
Lagertemperatur: Raumtemperatur
MDL: MFCD18391673
CAS-Nummer: 1261448-45-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR