4-Bromo-3-iodobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
OR400078-250MG
OR400078-1G
APOSOR400078-250MGEA
87
EUR
APOSOR400078-250MG
APOSOR400078-1G
4-Bromo-3-iodobenzyl bromide
4-Bromo-3-iodobenzyl bromide
Formel:
C₇H₅Br₂I Lagertemperatur: Raumtemperatur |
MDL:
MFCD18391673 CAS-Nummer: 1261448-45-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...