Methyl 2-bromo-5-hydroxybenzoate

Lieferant: Apollo Scientific
Achtung

OR400090-1G OR400090-5G OR400090-25G
APOSOR400090-1GEA 109 EUR
APOSOR400090-1G APOSOR400090-5G APOSOR400090-25G
Methyl 2-bromo-5-hydroxybenzoate
Methyl 2-bromo-5-hydroxybenzoate
Formel: C₈H₇BrO₃
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10566797
CAS-Nummer: 154607-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR